siRNA Details
| Gene Symbol: LOC101927151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110687.1 | 1 | 48 | |||
| NR_110690.1 | 1 | 48 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000586220.1 | - | 1 | 48 | Chr. 19 - 27793895 - 27798012 [+] on Build GRCh38 | |

LOC101927151
Genomic Map