siRNA Details
Gene Symbol: LINC00907 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046457.1 | 4, 5 | 501 | |||
GenBank mRNA | AB593075.1 | - | 4, 5 | 499 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000593051.5 | - | 4, 5 | 499 | Chr. 18 - 42186670 - 42462183 [+] on Build GRCh38 |
