siRNA Details
Gene Symbol: LINC01028 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104129.1 | 3 | 965 | |||
NR_104130.1 | 2 | 843 | ||||
GenBank mRNA | BX647864.1 | - | 2 | 838 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01028 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104129.1 | 3 | 965 | |||
NR_104130.1 | 2 | 843 | ||||
GenBank mRNA | BX647864.1 | - | 2 | 838 |