siRNA Details
| Gene Symbol: LOC101928844 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110740.1 | 4 | 1088 | |||
| GenBank mRNA | BC033124.1 | - | 4 | 1088 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000589276.1 | - | 4 | 1088 | Chr. 19 - 5847467 - 5858239 [+] on Build GRCh38 | |

LOC101928844
Genomic Map