siRNA Details
| Gene Symbol: LOC101928844 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110740.1 | 1 | 227 | |||
| GenBank mRNA | BC033124.1 | - | 1 | 227 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928844| Gene Symbol: LOC101928844 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110740.1 | 1 | 227 | |||
| GenBank mRNA | BC033124.1 | - | 1 | 227 | ||