siRNA Details
Gene Symbol: LINC01539 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040025.1 | 5 | 1000 | |||
NR_040026.1 | 3 | 533 | ||||
GenBank mRNA | AK057336.1 | - | 3 | 533 | ||
AK058169.1 | - | 5 | 981 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01539 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040025.1 | 5 | 1000 | |||
NR_040026.1 | 3 | 533 | ||||
GenBank mRNA | AK057336.1 | - | 3 | 533 | ||
AK058169.1 | - | 5 | 981 |