siRNA Details
| Gene Symbol: LINC01533 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110673.1 | 3 | 708 | |||
| NR_110674.1 | 3 | 552 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000588694.5 | - | 3 | 519 | Chr. 19 - 32025895 - 32039857 [+] on Build GRCh38 | |

LINC01533
Genomic Map