siRNA Details
| Gene Symbol: LOC100132174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104134.1 | 2 | 1774 | |||
| GenBank mRNA | AK127897.1 | - | 2 | 1774 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000593118.1 | - | 2 | 1774 | Chr. 17 - 77722872 - 77728559 [-] on Build GRCh38 | |

LOC100132174
Genomic Map