siRNA Details
| Gene Symbol: LOC146513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038438.1 | 3 | 1408 | |||
| GenBank mRNA | BC043214.2 | - | 3 | 1412 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000599486.1 | - | 3 | 1412 | Chr. 16 - 86286427 - 86293389 [+] on Build GRCh38 | |

LOC146513
Genomic Map