siRNA Details
| Gene Symbol: LOC101928295 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110729.1 | 3 | 2759 | |||
| GenBank mRNA | AK126060.1 | - | 3 | 2757 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000599433.1 | - | 3 | 2759 | Chr. 19 - 49368705 - 49388081 [-] on Build GRCh38 | |

LOC101928295
Genomic Map