siRNA Details
Gene Symbol: LOC284912 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC001801.1 | - | 1 | 111 | 3355513 | |
BG471420.1 | - | 1 | 112 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000472240.1 | - | 1 | 111 | Chr. 22 - 35626988 - 35635134 [-] on Build GRCh38 |
