siRNA Details
| Gene Symbol: LINC01082 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103859.1 | 2 | 395 | |||
| GenBank mRNA | BP423579.1 | - | 2 | 366 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000601250.1 | - | 2 | 395 | Chr. 16 - 86196181 - 86199720 [+] on Build GRCh38 | |

LINC01082
Genomic Map