siRNA Details
| Gene Symbol: ZNF350-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103847.1 | 2, 3 | 283 | |||
| GenBank mRNA | BC014606.2 | - | 2, 3 | 255 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF350-AS1| Gene Symbol: ZNF350-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103847.1 | 2, 3 | 283 | |||
| GenBank mRNA | BC014606.2 | - | 2, 3 | 255 | ||