siRNA Details
| Gene Symbol: ZSCAN16-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103455.1 | 3 | 2142 | |||
| NR_103456.1 | 2 | 1945 | ||||
| GenBank mRNA | AK127889.1 | - | 3 | 2120 | ||
| BC043177.2 | - | 2 | 1495 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000602810.1 | - | 2 | 1495 | Chr. 6 - 28121798 - 28136866 [-] on Build GRCh38 | |

ZSCAN16-AS1
Genomic Map