siRNA Details
| Gene Symbol: LINC01233 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110663.1 | 2 | 345 | |||
| NR_110664.1 | 1, 2 | 300 | ||||
| GenBank mRNA | BX092359.1 | - | 1, 2 | 300 | ||
| HY002653.1 | - | 1, 2 | 255 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000598832.1 | - | 1, 2 | 300 | Chr. 19 - 22532626 - 22533494 [+] on Build GRCh38 | |

LINC01233
Genomic Map