siRNA Details
Gene Symbol: LINC01233 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110663.1 | 2 | 539 | |||
NR_110664.1 | 2 | 494 | ||||
GenBank mRNA | HY002653.1 | - | 2 | 449 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000598832.1 | - | 2 | 494 | Chr. 19 - 22532626 - 22533494 [+] on Build GRCh38 |
