siRNA Details
Gene Symbol: BISPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130766.1 | 3 | 328 | |||
GenBank mRNA | AK055623.1 | - | 3 | 271 | ||
AK311380.1 | - | 3 | 545 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BISPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130766.1 | 3 | 328 | |||
GenBank mRNA | AK055623.1 | - | 3 | 271 | ||
AK311380.1 | - | 3 | 545 |