siRNA Details
| Gene Symbol: ZIM2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110744.1 | 4 | 1519 | |||
| GenBank mRNA | BC036412.1 | - | 4 | 1496 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000595954.1 | - | 4 | 1497 | Chr. 19 - 56765344 - 56799646 [+] on Build GRCh38 | |

ZIM2-AS1
Genomic Map