siRNA Details
| Gene Symbol: ZNF528-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125345.1 | 1 | 108 | |||
| GenBank mRNA | AK097590.1 | - | 1 | 116 | ||
| BI755704.1 | - | 1 | 117 | |||
| BX537909.1 | - | 1 | 83 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000598892.1 | - | 1 | 20 | Chr. 19 - 52396639 - 52397669 [-] on Build GRCh38 | |

ZNF528-AS1
Genomic Map