siRNA Details
| Gene Symbol: SPACA6P-AS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108100.1 | 2 | 2390 | |||
| GenBank mRNA | AK125996.1 | - | 2 | 2389 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000602324.1 | - | 2 | 2390 | Chr. 19 - 51685363 - 51693456 [-] on Build GRCh38 | |

SPACA6P-AS
Genomic Map