siRNA Details
| Gene Symbol: LOC440173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027471.1 | 2 | 828 | |||
| GenBank mRNA | BC027471.1 | - | 2 | 826 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000602579.1 | - | 2 | 828 | Chr. 9 - 87008455 - 87042126 [-] on Build GRCh38 | |

LOC440173
Genomic Map