siRNA Details
| Gene Symbol: LOC101928659 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135180.1 | 2 | 362 | |||
| GenBank mRNA | BQ640484.1 | - | 2 | 359 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000602282.2 | - | 2 | 362 | Chr. 16 - 87317509 - 87318037 [+] on Build GRCh38 | |

LOC101928659
Genomic Map