siRNA Details
| Gene Symbol: LOC101928557 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135193.1 | 2 | 307 | |||
| GenBank mRNA | BX118346.1 | - | 2 | 307 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928557| Gene Symbol: LOC101928557 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135193.1 | 2 | 307 | |||
| GenBank mRNA | BX118346.1 | - | 2 | 307 | ||