siRNA Details
| Gene Symbol: LOC101928557 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135193.1 | 2 | 192 | |||
| GenBank mRNA | BX118346.1 | - | 2 | 192 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000602617.1 | - | 2 | 192 | Chr. 16 - 85792415 - 85792933 [+] on Build GRCh38 | |

LOC101928557
Genomic Map