siRNA Details
Gene Symbol: SMC2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121580.1 | 3 | 985 | |||
GenBank mRNA | AK125887.1 | - | 3 | 986 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000603487.1 | - | 3 | 985 | Chr. 9 - 104080024 - 104092474 [-] on Build GRCh38 |
