siRNA Details
Gene Symbol: LINC01235 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033863.1 | 3 | 1887 | |||
GenBank mRNA | AK123194.1 | - | 3 | 1887 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000604724.5 | - | 3 | 1887 | Chr. 9 - 13406380 - 13431329 [-] on Build GRCh38 |
