siRNA Details
| Gene Symbol: LRRC37A8P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126038.1 | 1 | 250 | |||
| GenBank mRNA | AK128235.1 | - | 1 | 250 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000604907.1 | - | 1 | 250 | Chr. 17 - 35893707 - 35911023 [-] on Build GRCh38 | |

LRRC37A8P
Genomic Map