siRNA Details
Gene Symbol: LRRC37A8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126038.1 | 1 | 3474 | |||
GenBank mRNA | AK125932.1 | - | 1 | 2400 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000604907.1 | - | 1 | 3474 | Chr. 17 - 35893707 - 35911023 [-] on Build GRCh38 |
