siRNA Details
| Gene Symbol: NAMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102270.1 | 10 | 2134 | |||
| NR_102271.1 | 6 | 1582 | ||||
| GenBank mRNA | DQ673922.1 | - | 10 | 2103 | ||
| DQ673929.1 | - | 2 | 238 | |||
| DQ673934.1 | - | 3 | 363 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NAMA| Gene Symbol: NAMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102270.1 | 10 | 2134 | |||
| NR_102271.1 | 6 | 1582 | ||||
| GenBank mRNA | DQ673922.1 | - | 10 | 2103 | ||
| DQ673929.1 | - | 2 | 238 | |||
| DQ673934.1 | - | 3 | 363 | |||