siRNA Details
| Gene Symbol: NAMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102270.1 | 3 | 589 | |||
| NR_102271.1 | 3 | 589 | ||||
| GenBank mRNA | DQ673922.1 | - | 3 | 559 | ||
| DQ673923.1 | - | 3 | 559 | |||
| DQ673924.1 | - | 3 | 559 | |||
| DQ673926.1 | - | 3 | 559 | |||
| DQ673936.1 | - | 2 | 116 | |||
| DQ673946.1 | - | 1 | 1123 | |||
| DQ673949.1 | - | 2 | 444 | |||
| DQ673950.1 | - | 2 | 221 | |||
| DQ673953.1 | - | 2 | 382 | |||
| DQ673965.1 | - | 4 | 449 | |||

NAMA
Genomic Map