siRNA Details
| Gene Symbol: LOC101927811 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110119.1 | 4 | 444 | |||
| NR_110120.1 | 3 | 351 | ||||
| GenBank mRNA | AV707592.1 | - | 4 | 376 | ||
| HY150107.1 | - | 3 | 352 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927811| Gene Symbol: LOC101927811 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110119.1 | 4 | 444 | |||
| NR_110120.1 | 3 | 351 | ||||
| GenBank mRNA | AV707592.1 | - | 4 | 376 | ||
| HY150107.1 | - | 3 | 352 | |||