siRNA Details
| Gene Symbol: ARMCX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK023444.1 | - | 1 | 1021 | ||
| BC050678.1 | - | 6 | 1336 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000476910.6 | - | 6 | 1336 | Chr. X - 102599537 - 102640507 [+] on Build GRCh38 | |

ARMCX5
Genomic Map