siRNA Details
| Gene Symbol: LINC00466 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038252.2 | 12 | 1693 | |||
| GenBank mRNA | BC040309.1 | - | 12 | 1666 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00466| Gene Symbol: LINC00466 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038252.2 | 12 | 1693 | |||
| GenBank mRNA | BC040309.1 | - | 12 | 1666 | ||