siRNA Details
Gene Symbol: CYB561D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_111912.1 | 4 | 855 | |||
NR_111913.1 | 3 | 937 | ||||
NR_111914.1 | 3 | 918 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CYB561D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_111912.1 | 4 | 855 | |||
NR_111913.1 | 3 | 937 | ||||
NR_111914.1 | 3 | 918 |