siRNA Details
Gene Symbol: CYB561D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_111913.1 | 1 | 316 | |||
NR_111914.1 | 1 | 316 | ||||
GenBank mRNA | BM704874.1 | - | 1 | 316 | ||
CT000022.1 | - | 1 | 265 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CYB561D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_111913.1 | 1 | 316 | |||
NR_111914.1 | 1 | 316 | ||||
GenBank mRNA | BM704874.1 | - | 1 | 316 | ||
CT000022.1 | - | 1 | 265 |