siRNA Details
Gene Symbol: AGAP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133447.1 | NP_597704.1 | 1, 2 | 183 | ||
GenBank mRNA | AK097165.1 | - | 1, 2 | 183 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000433214.2 | - | 1, 2 | 183 | Chr. 10 - 86970741 - 87010126 [+] on Build GRCh38 |
