siRNA Details
| Gene Symbol: ZNF165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AL583605.3 | - | 1, 2 | 514 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000605945.1 | - | 1, 2 | 514 | Chr. 6 - 28078792 - 28081130 [-] on Build GRCh38 | |

ZNF165
Genomic Map