siRNA Details
Gene Symbol: LINC01607 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125410.1 | 2 | 105 | |||
GenBank mRNA | AI801879.1 | - | 2 | 106 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000607172.1 | - | 2 | 105 | Chr. 8 - 79768142 - 79802842 [+] on Build GRCh38 |
