siRNA Details
| Gene Symbol: LYRM4-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126016.1 | 3 | 943 | |||
| NR_126017.1 | 3 | 861 | ||||
| NR_126018.1 | 2 | 848 | ||||
| NR_126019.1 | 2 | 930 | ||||
| GenBank mRNA | BC042032.1 | - | 2 | 848 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000606761.1 | - | 2 | 848 | Chr. 6 - 5029972 - 5043449 [+] on Build GRCh38 | |

LYRM4-AS1
Genomic Map