siRNA Details
| Gene Symbol: LYRM4-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126018.1 | 1 | 145 | |||
| NR_126019.1 | 1 | 145 | ||||
| GenBank mRNA | BC042032.1 | - | 1 | 145 | ||
| BC133670.1 | - | 1 | 86 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LYRM4-AS1| Gene Symbol: LYRM4-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126018.1 | 1 | 145 | |||
| NR_126019.1 | 1 | 145 | ||||
| GenBank mRNA | BC042032.1 | - | 1 | 145 | ||
| BC133670.1 | - | 1 | 86 | |||