siRNA Details
| Gene Symbol: ARF4-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132369.1 | 3 | 537 | |||
| NR_132370.1 | 2 | 426 | ||||
| GenBank mRNA | BF793425.1 | - | 3 | 530 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ARF4-AS1| Gene Symbol: ARF4-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132369.1 | 3 | 537 | |||
| NR_132370.1 | 2 | 426 | ||||
| GenBank mRNA | BF793425.1 | - | 3 | 530 | ||