siRNA Details
| Gene Symbol: ARF4-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132369.1 | 1 | 245 | |||
| NR_132370.1 | 1 | 245 | ||||
| GenBank mRNA | BF793425.1 | - | 1 | 238 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000607297.1 | - | 1 | 245 | Chr. 3 - 57597789 - 57600916 [+] on Build GRCh38 | |

ARF4-AS1
Genomic Map