siRNA Details
| Gene Symbol: LOC101929352 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134478.1 | 3 | 826 | |||
| GenBank mRNA | AK125507.1 | - | 2 | 1785 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000605929.1 | - | 2 | 1784 | Chr. 10 - 31187883 - 31261910 [+] on Build GRCh38 | |

LOC101929352
Genomic Map