siRNA Details
| Gene Symbol: FAM231D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_111934.1 | 1, 2 | 1447 | |||
| Gene Symbol: LOC388692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_111933.1 | 1, 2 | 1768 | |||

FAM231D
Genomic Map