siRNA Details
Gene Symbol: LOC401286 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_117091.1 | 3 | 4502 | |||
GenBank mRNA | AK127120.1 | - | 3 | 4502 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000609107.1 | - | 3 | 4549 | Chr. 6 - 167666840 - 167679270 [-] on Build GRCh38 |
