siRNA Details
| Gene Symbol: AFAP1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026892.1 | 2 | 3892 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000608442.1 | - | 2 | 3892 | Chr. 4 - 7754090 - 7778928 [+] on Build GRCh38 | |

AFAP1-AS1
Genomic Map