siRNA Details
| Gene Symbol: LOC101929234 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110304.1 | 3 | 962 | |||
| GenBank mRNA | AK307176.1 | - | 3 | 962 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000609200.1 | - | 3 | 962 | Chr. 10 - 75409157 - 75411842 [+] on Build GRCh38 | |

LOC101929234
Genomic Map