siRNA Details
Gene Symbol: LOC102723582 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125405.1 | 2 | 57 | |||
GenBank mRNA | DA501747.1 | - | 2 | 57 | ||
HY006567.1 | - | 2 | 56 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000608346.1 | - | 2 | 44 | Chr. 3 - 122416219 - 122433262 [+] on Build GRCh38 |
