siRNA Details
| Gene Symbol: ALDH1A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK056534.1 | - | 5 | 713 | ||
| Gene Symbol: LOC105369201 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011522309.2 | XP_011520611.1 | 1 | 809 | ||
| XM_011522311.2 | XP_011520613.1 | 1 | 809 | |||
| XM_011522312.2 | XP_011520614.1 | 1 | 809 | |||
| XM_011522313.2 | XP_011520615.1 | 1 | 809 | |||
| XM_017022802.1 | XP_016878291.1 | 1 | 809 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000431060.4 | - | 5 | 713 | Chr. 15 - 100849831 - 100864296 [+] on Build GRCh38 | |

ALDH1A3
Genomic Map