siRNA Details
Gene Symbol: LOC101927391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110084.1 | 4 | 713 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000609497.5 | - | 3 | 1556 | Chr. 7 - 7552462 - 7566996 [-] on Build GRCh38 |
